Merck
  • Sohlh2 inhibits the apoptosis of mouse primordial follicle oocytes via C-kit/PI3K/Akt/Foxo3a signalling pathway.
Sohlh2 inhibits the apoptosis of mouse primordial follicle oocytes via C-kit/PI3K/Akt/Foxo3a signalling pathway.
Reproductive biomedicine online (2015-03-17)
Xiaoli Zhang, Haiyu Zhang, Qing Gao, Shufang Ji, Lujun Bing, Jing Hao
ABSTRACT
We previously reported that bone morphogenetic protein 4/drosophila mothers against decapentaplegic protein (BMP4/Smad) signalling pathway initiated primordial follicle growth and prevented oocyte apoptosis via up-regulation of Sohlh2 and receptor for kit ligand (c-kit). The mechanism underlying this process was not fully elucidated. In the present study, primary oocyte cultures were established from ovaries of 3-day-old female mouse pups by two-step enzyme digestion. Cultures were divided into Sohlh2 small interference RNA (SiRNA) group, negative SiRNA group, Sohlh2 overexpression plasmid group and pCAG-puro group. TdT (terminal deoxynucleotidyl transferase)-mediated dUDP nick-end labelling assay was carried out to detect the oocyte apoptosis; immunocytochemical staining and quantitative real time-polymerase chain reaction detected the expression of c-kit and Forkhead box O3a (Foxo3a); Western blot was performed to detect the expression of Sohlh2, C-kit, saerine/threonine kinases (Akt1) and Foxo3a. The results showed that Sohlh2 inhibited oocyte apoptosis and upregulated c-kit expression; Sohlh2 decreased the endonuclear Foxo3a via the upregulation of phosphorylated Akt1 (P-Akt1) and phosphorylated Foxo3a (P-Foxo3a) but not total Akt1 (T-Akt1) or total Foxo3a (T-Foxo3a); Sohlh2 increased P-Akt1 but not T-Akt1; the PI3K (phosphotidylinsitol-3-kinase) inhibitor LY294002 ameliorated the role of Sohlh2 on phosphorylation of Akt1 and Foxo3a. Sohlh2 may inhibit oocyte apoptosis via c-kit/PI3K/Akt/Foxo3a signalling pathway.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis