Merck
  • Stretchable Heater Using Ligand-Exchanged Silver Nanowire Nanocomposite for Wearable Articular Thermotherapy.
Stretchable Heater Using Ligand-Exchanged Silver Nanowire Nanocomposite for Wearable Articular Thermotherapy.
ACS nano (2015-06-02)
Suji Choi, Jinkyung Park, Wonji Hyun, Jangwon Kim, Jaemin Kim, Young Bum Lee, Changyeong Song, Hye Jin Hwang, Ji Hoon Kim, Taeghwan Hyeon, Dae-Hyeong Kim
ABSTRACT
Thermal therapy is one of the most popular physiotherapies and it is particularly useful for treating joint injuries. Conventional devices adapted for thermal therapy including heat packs and wraps have often caused discomfort to their wearers because of their rigidity and heavy weight. In our study, we developed a soft, thin, and stretchable heater by using a nanocomposite of silver nanowires and a thermoplastic elastomer. A ligand exchange reaction enabled the formation of a highly conductive and homogeneous nanocomposite. By patterning the nanocomposite with serpentine-mesh structures, conformal lamination of devices on curvilinear joints and effective heat transfer even during motion were achieved. The combination of homogeneous conductive elastomer, stretchable design, and a custom-designed electronic band created a novel wearable system for long-term, continuous articular thermotherapy.
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate, ≥99.999% trace metals basis
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Copper(II) chloride, powder, 99%
Sigma-Aldrich
Copper(II) chloride, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Copper(II) chloride, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) chloride, 97%