Merck
  • Alteration of O-GlcNAcylation affects serine phosphorylation and regulates gene expression and activity of pyruvate kinase M2 in colorectal cancer cells.
Alteration of O-GlcNAcylation affects serine phosphorylation and regulates gene expression and activity of pyruvate kinase M2 in colorectal cancer cells.
Oncology reports (2015-08-08)
Parunya Chaiyawat, Daranee Chokchaichamnankit, Kriengsak Lirdprapamongkol, Chantragan Srisomsap, Jisnuson Svasti, Voraratt Champattanachai
ABSTRACT
O-GlcNAcylation is a dynamic post-translational modification that has extensive crosstalk with phosphorylation either at the same or adjacent sites of various proteins. We have previously reported that O-GlcNAcylation level was increased in primary breast and colorectal cancer, but the interplay of the two modifications remains unclear. Therefore, we explored crosstalk of the modifications by RNA interference against O-GlcNAc transferase (OGT) in colorectal cancer cells. Two-dimensional immunoblotting and mass spectrometric analysis showed that the levels of O-GlcNAc and serine phosphorylation of many proteins including serine hydroxymethyltransferase, cytokeratin-8, pyruvate kinase M2 (PKM2), heterogeneous nuclear ribonucleoprotein L, and lamin-B1, were reduced in siOGT cells compared to siScramble cells. In HT29 cells, immunoprecipitated PKM2 revealed decreased O-GlcNAc and serine phosphorylation levels after siOGT knockdown, but increased levels after treatment with Thiamet-G, an inhibitor of O-GlcNAcase (OGA). In addition, when global O-GlcNAcylation was enhanced by treating cells with Thiamet-G, PKM2 expression level was upregulated, but PKM2-specific activity was decreased. On the other hand, in OGT knockdown cells, PKM2 expression level was downregulated, but PKM2-specific activity was increased. Moreover, the metastatic colorectal cancer cells, SW620, had more O-GlcNAc-PKM2 and showed lower PKM2-specific activity compared to the non-metastatic colorectal cancer SW480 cells. These results suggested roles of O-GlcNAcylation in modulating serine phosphorylation, as well as in regulating PKM2 activity and expression. Interfering levels of O-GlcNAcylation of PKM2 may be a novel target in controlling cancer metabolism and tumorigenesis of colorectal cancer.
Sigma-Aldrich
Trimethylgallium, packaged for use in deposition systems
Sigma-Aldrich
Methyl-β-D-thiogalactoside
Sigma-Aldrich
O-Phospho-L-serine
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
MISSION® esiRNA, targeting human OGT
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ogt
SAFC
Iodoacetamide
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis