Merck
  • Limonoids from the Seeds of Swietenia macrophylla and Their Anti-Inflammatory Activities.
Limonoids from the Seeds of Swietenia macrophylla and Their Anti-Inflammatory Activities.
Molecules (Basel, Switzerland) (2015-10-17)
Li-Chai Chen, Hsiang-Ruei Liao, Pei-Yu Chen, Wen-Lung Kuo, Tsung-Hsien Chang, Ping-Jyun Sung, Zhi-Hong Wen, Jih-Jung Chen
ABSTRACT
A new limonoid, swietemacrophin (1), was isolated from the seeds of Swietenia macrophylla, together with five known compounds 2-6. The structure of 1 was determined through extensive 1D/2D-NMR and mass-spectrometric analyses. Swietemacrophin (1), humilinolide F (2), 3,6-O,O-diacetylswietenolide (3), 3-O-tigloylswietenolide (4), and swietemahonin E (5) exhibited inhibition (IC50 values≤45.44 μM) of superoxide anion generation by human neutrophils in response to formyl-L-methionyl-L-leucyl-L-phenylalanine (fMLP). Compounds 1, 4, 5, and swietenine (6) showed potent inhibition with IC50 values≤36.32 μM, against lipopolysaccharide (LPS)-induced nitric oxide (NO) generation.
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium bromide, ≥99% trace metals basis
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%