Merck
  • A Simple Chiral Cu(II) Complex as an Effective Phase-Transfer Catalyst for the Enantioselective Alkylation of Dissymmetric Glycinate Ketimines.
A Simple Chiral Cu(II) Complex as an Effective Phase-Transfer Catalyst for the Enantioselective Alkylation of Dissymmetric Glycinate Ketimines.
Chirality (2015-10-03)
Hanane Bafqiren, Hafid Zouihri, Said Gmouh, Jamal Jamal Eddine
ABSTRACT
Catalytic asymmetric benzylation of a dissymmetric tert-butylglycinate ketimine, incorporating 1-naphthyl and phenyl groups as the Schiff base substituents, under phase-transfer conditions was investigated. It was interesting to note that the sense of asymmetric induction of the alkylation of Z-imine stereoisomer is opposite to that of the corresponding E stereoisomer with a similar degree of enantioselectivity. More interestingly, the chiral Cu(II) complex of the Schiff base derived from (R)-2-phenylglycinol and 2-hydroxy-1-naphthaldehyde was found to catalyze the same reaction under solid-liquid conditions with comparable enantioselectivity (up to 60% ee) with respect to known cinchona alkaloid catalysts. The solvent/base-system parameter was shown to control the optimal catalytic activity.
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS