Merck
Lipid Metabolites from the Mushroom Meripilus giganteus.
Natural product communications (2016-01-12)
Francesca Catenia, Tiziano Altieri, Marina Zacchigna, Giuseppe Procida, Jelena Zilic, Dusan Zigon, Angelo Cichelli
ABSTRACT
The phytochemical investigation of the methanolic extract of the white rot fungus Meripilus giganteus resulted in the isolation and identification of complex mixtures of free fatty acids (1), monoacylglycerols (2), cerebrosides (3), ergosterol (4) and ergosterol peroxide (5). The structures of the isolated lipid metabolites (1-5) were determined by chemical and spectroscopic methods. The antioxidant activity of the whole MeOH extract of the fungus was evaluated through in vitro model systems, such as 2,2-diphenyl-l-picrylhydrazyl (DPPH) and superoxide anion. In all two systems, the results indicated that the extract of the fungus showed the same free-radical-scavenging activity with SC50 data of 47.70 µg/mL, compared with the positive control quercetin (DPPH assay). None of the isolated compounds (1-5) showed a significant activity. Compounds 2-4 were isolated from Meripilus giganteus for the first time.
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%