Merck
  • Synthesis of a novel ionic liquid modified copolymer hydrogel and its rapid removal of Cr (VI) from aqueous solution.
Synthesis of a novel ionic liquid modified copolymer hydrogel and its rapid removal of Cr (VI) from aqueous solution.
Journal of colloid and interface science (2015-06-10)
Yinhua Jiang, Fan Li, Guibing Ding, Yecheng Chen, Yan Liu, Yuanzhi Hong, Peipei Liu, Xiuxiu Qi, Liang Ni
ABSTRACT
A novel ionic liquid modified copolymer hydrogel (PAMDA) was successfully synthesized by a simple water solution copolymerization using acrylamide (AM), dimethyldiallylammonium chloride (DADMAC) and ionic liquid (1-allyl-3-methylimidazolium chloride; [Amim]Cl) as copolymerization monomers. The structure and morphology of as-prepared copolymer hydrogel PAMDA were confirmed by Fourier transform infrared (FT-IR), field-emission scanning electron microscope (FE-SEM) and thermogravimetric analysis (TG). The copolymer hydrogel was applied as a novel adsorbent for the rapid removal of Cr (VI) from aqueous solution. The effects of several parameters such as the content of ionic liquid [Amim]Cl, solution pH, contact time, adsorbent dosage and initial Cr (VI) concentration on the adsorption were also investigated. The modification of [Amim]Cl significantly enhanced Cr (VI) adsorption. The adsorption equilibrium data fitted with Langmuir isotherm model better than Freundlich isotherm model. The maximum adsorption capacity for Cr (VI) ions was 74.5 mg L(-1) at 323 K based on Langmuir isotherm model. The removal rate could reach 95.9% within 10 min at 323 K and the adsorption process of Cr (VI) on PAMDA was well described by the pseudo-second-order kinetic model. The activation energy of adsorption was further investigated and found to be 1.094 kJ mol(-1), indicating the adsorption of Cr (VI) onto PAMDA was physisorption.
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
1,5-Diphenylcarbazide, reagent grade
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), 98%
Sigma-Aldrich
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, Molecular Biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
1,5-Diphenylcarbazide, ACS reagent
Sigma-Aldrich
N,N′-Methylenebisacrylamide solution, suitable for electrophoresis, 2% in H2O
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%