Merck
  • Co-administration of deflazacort and doxycycline: a potential pharmacotherapy for Duchenne muscular dystrophy.
Co-administration of deflazacort and doxycycline: a potential pharmacotherapy for Duchenne muscular dystrophy.
Clinical and experimental pharmacology & physiology (2015-05-12)
Juliano Alves Pereira, Maria Julia Marques, Humberto Santo Neto
ABSTRACT
The standard therapy used in the treatment of Duchenne muscle dystrophy (DMD) is corticoids, such as deflazacort and prednisone. However, they have limited therapeutic value, and their combination with drugs already in use to treat other human diseases could potentially increase corticoid outcomes in DMD. In the present study, we evaluated whether a combined therapy of the corticoid deflazacort with doxycycline could result in greater improvement in mdx dystrophy than deflazacort alone. Deflazacort alone or deflazacort/doxycycline were administered for 36 days (starting on postnatal day 0) in drinking water. Histopathological, biochemical (creatine kinase), functional (forelimb muscle grip strength and fatigue) parameters and inflammatory markers (MMP-9, TNF-α, NF-kB) were evaluated in biceps brachii and diaphragm muscles of the mdx mice. The combined therapy was superior in improving the dystrophic phenotype compared to monotherapy. The primary results were observed in attenuating muscle fatigue, decreasing muscle total calcium and inflammatory markers and increasing β-dystroglycan, a main component of the dystrophin-protein complex. Furthermore, the combined therapy was effective in preventing the loss of body mass observed with deflazacort alone at this very early stage of therapy. The present study offers preclinical data to support further studies with deflazacort/doxycycline combined therapy in DMD clinical trials.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Dabco® 33-LV
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Xylazine hydrochloride, ≥99.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O