Merck
  • Chelerythrine-lysozyme interaction: spectroscopic studies, thermodynamics and molecular modeling exploration.
Chelerythrine-lysozyme interaction: spectroscopic studies, thermodynamics and molecular modeling exploration.
Physical chemistry chemical physics : PCCP (2015-06-05)
Chandrima Jash, Pritha Basu, Pavan V Payghan, Nanda Ghoshal, Gopinatha Suresh Kumar
ABSTRACT
The binding of the iminium and alkanolamine forms of chelerythrine to lysozyme (Lyz) was investigated by spectroscopy and docking studies. The thermodynamics of the binding was studied by calorimetry. Spectroscopic evidence suggested that Trp-62 and Trp-63 in the β-domain of the protein are closer to the binding site; moreover, the binding site was at a distance of 2.27 and 2.00 nm from the iminium and alkanolamine forms, respectively, according to the Forster theory of non-radiation energy transfer. The equilibrium binding constants for the iminium and alkanolamine forms at 298 K were evaluated to be 1.29 × 10(5) and 7.79 × 10(5) M(-1), respectively. The binding resulted in an alteration of the secondary structure of the protein with a distinct reduction of the helical organization. The binding of iminium was endothermic, involving electrostatic and hydrophobic interactions, while that of alkanolamine form was exothermic and dominated by hydrogen bonding interactions. Docking studies provided the atomistic details pertaining to the binding of both forms of chelerythrine and supported the higher binding in favour of the alkanolamine over the iminium. Furthermore, molecular dynamics study provided accurate insights regarding the binding of both chelerythrine forms in accordance with the experimental results obtained. Chelerythrine binding pocket involves the catalytic region and aggregation prone K-peptide region, which are sandwiched between one another. Overall, these results suggest that both the forms of the alkaloid bind to the protein but the neutral form has higher affinity than the cationic form.
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
Indole, ≥99%
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
Indole, ≥99%, FG