Merck
  • Fabrication of doxorubicin-loaded ellipsoid micelle based on diblock copolymer with a linkage of enzyme-cleavable peptide.
Fabrication of doxorubicin-loaded ellipsoid micelle based on diblock copolymer with a linkage of enzyme-cleavable peptide.
Colloids and surfaces. B, Biointerfaces (2015-07-06)
Haitao Huang, Jianqi Geng, Jafar Golzarian, Jin Huang, Jiahui Yu
ABSTRACT
In this study, a novel micellar drug carrier was fabricated from an amphiphilic diblock copolymer containing poly (ethylene glycol) monomethyl ether (mPEG) and stearic moiety (C18) with a linkage of valine-citrulline (VC), which can be cleaved by cathepsin B (CB) enriched in lysosome of tumor cell. Moreover, the self-assembled micelles were observed as ellipsoid shape with major and minor axis of ca. 169 and 103nm, respectively. Such drug carrier was used to encapsulate anti-cancer drug doxorubicin (DOX), and hence showed a faster drug release behavior in a mimic lysosome condition containing cathepsin B. It was ascribed to the lysosome-sensitivity of the valine-citrulline linkage, which was verified by the size distribution curve shifted to greater size under the same mimic lysosome condition. Furthermore, in comparison with pristine doxorubicin, the encapsulation strategy of as-fabricated micellar carrier resulted in a predominant decrease of cytotoxicity. On the whole, a micellar drug carrier, which can be disassembled by cathepsin B, has been emerging as a potential of specific drug release in lysosome. Additionally, the controlled nanoscale together with elongated structure of such assembled ellipsoid micelles might contribute passive targeting function to tumor tissue and faster cellular uptake behavior (Zhou et al.).
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
1,2-Dimethoxyethane, anhydrous, 99.5%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Piperidine, ReagentPlus®, 99%
Sigma-Aldrich
Piperidine, ≥99.5%, purified by redistillation
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Dimethyl ether, ≥99.9%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate