Merck
  • Toward the stereochemical identification of prohibited characterizing flavors in tobacco products: the case of strawberry flavor.
Toward the stereochemical identification of prohibited characterizing flavors in tobacco products: the case of strawberry flavor.
Archives of toxicology (2015-07-04)
Meike Paschke, Christoph Hutzler, Frank Henkler, Andreas Luch
ABSTRACT
With the revision of the European Tobacco Products Directive (2014/40/EU), characterizing flavors such as strawberry, candy, vanillin or chocolate will be prohibited in cigarettes and fine-cut tobacco. Product surveillance will therefore require analytical means to define and subsequently detect selected characterizing flavors that are formed by supplemented flavors within the complex matrix tobacco. We have analyzed strawberry-flavored tobacco products as an example for characterizing fruit-like aroma. Using this approach, we looked into aroma components to find indicative patterns or features that can be used to satisfy obligatory product information as requested by the European Directive. Accordingly, a headspace solid-phase microextraction (HS-SPME) technique was developed and coupled to subsequent gas chromatography-mass spectrometry (GC/MS) to characterize different strawberry-flavored tobacco products (cigarettes, fine-cut tobacco, liquids for electronic cigarettes, snus, shisha tobacco) for their volatile additives. The results were compared with non-flavored, blend characteristic flavored and other fruity-flavored cigarettes, as well as fresh and dried strawberries. Besides different esters and aldehydes, the terpenes linalool, α-terpineol, nerolidol and limonene as well as the lactones γ-decalactone, γ-dodecalactone and γ-undecalactone could be verified as compounds sufficient to convey some sort of strawberry flavor to tobacco. Selected flavors, i.e., limonene, linalool, α-terpineol, citronellol, carvone and γ-decalactone, were analyzed further with respect to their stereoisomeric composition by using enantioselective HS-SPME-GC/MS. These experiments confirmed that individual enantiomers that differ in taste or physiological properties can be distinguished within the tobacco matrix. By comparing the enantiomeric composition of these compounds in the tobacco with that of fresh and dried strawberries, it can be concluded that non-natural strawberry aroma is usually used to produce strawberry-flavored tobacco products. Such authenticity control can become of interest particularly when manufacturers claim that natural sources were used for flavoring of products. Although the definition of characterizing flavors by analytical means remains challenging, specific compounds or features are required to be defined for routine screening of reported information. Clarifications by sensory testing might still be necessary, but could be limited to a few preselected samples.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
SPME Fiber Assembly, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Manual Holder, pk of 3, purple hub
Supelco
SPME Fiber Assembly, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 3, purple hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, white hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, red hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, green hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, white hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, white hub
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, green hub
Supelco
SPME Portable Field Sampler, coating PDMS
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Linalool, ≥97%, FCC, FG
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, pink hub