Merck
  • Direct Derivatization vs Aqueous Extraction Methods of Fecal Free Fatty Acids for GC-MS Analysis.
Direct Derivatization vs Aqueous Extraction Methods of Fecal Free Fatty Acids for GC-MS Analysis.
Lipids (2015-05-27)
Bashar Amer, Caroline Nebel, Hanne C Bertram, Grith Mortensen, Trine K Dalsgaard
ABSTRACT
A comprehensive and accurate determination of free fatty acids (FFA) is required for fecal metabolomic investigations. The present study compares three aqueous extraction methods (1) ULTRA-TURRAX(®), (2) whirl mixing and (3) basic ULTRA-TURRAX extraction of fecal FFA with a direct derivatization approach using ethyl chloroformate as the derivatization reagent before determination by gas chromatography-mass spectrometry. The direct derivatization method resulted in significantly higher estimations (P < 0.01) of short- and long-chain fatty acids than was the case when applying the aqueous extraction methods using ULTRA-TURRAX, whirl mixing, or basic ULTRA-TURRAX extraction before the derivatization step. Thus, avoiding an aqueous extraction before derivatization reduces the loss of volatile short-chain FFA and the less water-soluble long-chain FFA.
Sigma-Aldrich
Myristic acid, ≥95%, FCC, FG
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Linolenic acid, ≥99%
Sigma-Aldrich
Decanoic acid, ≥98.0%
Sigma-Aldrich
Nonanoic acid, ≥97%
Sigma-Aldrich
Octanoic acid, ≥99%
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Octanoic acid, ≥98%
Sigma-Aldrich
Hexanoic acid, ≥98%, FCC, FG
Sigma-Aldrich
Octanoic acid, natural, ≥98%, FG
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
2-Methylbutyric acid, ≥98%, FG
Sigma-Aldrich
Myristic acid, natural, ≥98.5%, FG
Sigma-Aldrich
Nonanoic acid, 96%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Decanoic acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Decanoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Linolenic acid, ~70% (GC)