Merck
  • Brain-derived neurotrophic factor signaling does not stimulate subventricular zone neurogenesis in adult mice and rats.
Brain-derived neurotrophic factor signaling does not stimulate subventricular zone neurogenesis in adult mice and rats.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2008-12-17)
Rui P Galvão, José Manuel Garcia-Verdugo, Arturo Alvarez-Buylla
ABSTRACT
In rodents, the adult subventricular zone (SVZ) generates neuroblasts which migrate to the olfactory bulb (OB) and differentiate into interneurons. Recent work suggests that the neurotrophin Brain-Derived Neurotrophic Factor (BDNF) can enhance adult SVZ neurogenesis, but the mechanism by which it acts is unknown. Here, we analyzed the role of BDNF and its receptor TrkB in adult SVZ neurogenesis. We found that TrkB is the most prominent neurotrophin receptor in the mouse SVZ, but only the truncated, kinase-negative isoform (TrkB-TR) was detected. TrkB-TR is expressed in SVZ astrocytes and ependymal cells, but not in neuroblasts. TrkB mutants have reduced SVZ proliferation and survival and fewer new OB neurons. To test whether this effect is cell-autonomous, we grafted SVZ cells from TrkB knock-out mice (TrkB-KO) into the SVZ of wild-type mice (WT). Grafted progenitors generated neuroblasts that migrated to the OB in the absence of TrkB. The survival and differentiation of granular interneurons and Calbindin(+) periglomerular interneurons seemed unaffected by the loss of TrkB, whereas dopaminergic periglomerular neurons were reduced. Intra-ventricular infusion of BDNF yielded different results depending on the animal species, having no effect on neuron production from mouse SVZ, while decreasing it in rats. Interestingly, mice and rats also differ in their expression of the neurotrophin receptor p75. Our results indicate that TrkB is not essential for adult SVZ neurogenesis and do not support the current view that delivering BDNF to the SVZ can enhance adult neurogenesis.
Sigma-Aldrich
Anti-Trk B Antibody, pain, a.a. 54-67 rTrkB, serum, Chemicon®
Sigma-Aldrich
Anti-Nerve Growth Factor Receptor Antibody, extracellular, clone 192-IgG, clone 192-IgG, Chemicon®, from mouse
Sigma-Aldrich
Anti-TrkB Antibody, serum, Chemicon®
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, clone GA5, Chemicon®, from mouse
Roche
In Situ Cell Death Detection Kit, TMR red, sufficient for ≤50 tests
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit