Merck
One-way PCR-based mapping of a replication initiation point (RIP).
Nature protocols (2008-10-18)
Julia Romero, Hoyun Lee
ABSTRACT
The lamin B2 locus is the only mammalian origin whose replication initiation points (RIPs) have been mapped. Although this paper was published 8 years ago, no further mammalian RIP-mapping studies have been reported, largely due to technical difficulties of ligation-mediated (LM)-PCR used by the authors. Here, we report the development of a simple, one-way PCR-based protocol that allows one to accurately determine RIPs at mammalian origins. The procedure can be completed within 48 h from the time of cell lysis in the agarose gel. Nascent DNA is then isolated from the same gel after DNA is separated by alkaline gel electrophoresis. Subsequently, RIPs are determined by one-way PCR-based primer extension using labeled primers. Using this protocol, we have successfully mapped RIPs in the human DBF4 locus. As one-way PCR is routinely used by many scientists, this protocol will provide a powerful new tool for studying DNA replication in many organisms including mammalian cells.
Product Description
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, Molecular Biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Emetine dihydrochloride
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, Molecular Biology, ≥99% (GC)