Merck
  • Denhaminols A-H, dihydro-β-agarofurans from the endemic Australian rainforest plant Denhamia celastroides.
Denhaminols A-H, dihydro-β-agarofurans from the endemic Australian rainforest plant Denhamia celastroides.
Journal of natural products (2015-01-13)
Claire Levrier, Martin C Sadowski, Colleen C Nelson, Peter C Healy, Rohan A Davis
ABSTRACT
Eight new dihydro-β-agarofurans, denhaminols A-H (1-8), were isolated from the leaves of the Australian rainforest tree Denhamia celastroides. The chemical structures of 1-8 were elucidated following analysis of 1D/2D NMR and MS data. The absolute configuration of denhaminol A (1) was determined by single-crystal X-ray crystallography. All compounds were evaluated for cytotoxic activity against the human prostate cancer cell line LNCaP, using live-cell imaging and metabolic assays. Denhaminols A (1) and G (7) were also tested for their effects on the lipid content of LNCaP cells. This is the first report of secondary metabolites from D. celastroides.
Sigma-Aldrich
Nile Red, BioReagent, suitable for fluorescence, ≥97.0% (HPLC)