Merck
  • Comparative cytotoxicity of chelidonine and homochelidonine, the dimethoxy analogues isolated from Chelidonium majus L. (Papaveraceae), against human leukemic and lung carcinoma cells.
Comparative cytotoxicity of chelidonine and homochelidonine, the dimethoxy analogues isolated from Chelidonium majus L. (Papaveraceae), against human leukemic and lung carcinoma cells.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2016-03-13)
Radim Havelek, Martina Seifrtova, Karel Kralovec, Eliska Krocova, Veronika Tejkalova, Ivan Novotny, Lucie Cahlikova, Marcela Safratova, Lubomir Opletal, Zuzana Bilkova, Jirina Vavrova, Martina Rezacova
ABSTRACT
The search for new anticancer compounds is a crucial element of natural products research. In this study the effects of naturally occurring homochelidonine in comparison to chelidonine on cell cycle progression and cell death in leukemic T-cells with different p53 status are described. The mechanism of cytotoxic, antiproliferative, apoptosis-inducing effects and the effect on expressions of cell cycle regulatory proteins was investigated using XTT assay, Trypan blue exclusion assay, flow cytometry, Western blot analysis, xCELLigence, epi-fluorescence and 3D super resolution microscopy. A549 cells were used for xCELLigence, clonogenic assay and for monitoring microtubule stability. We found that homochelidonine and chelidonine displayed significant cytotoxicity in examined blood cancer cells with the exception of HEL 92.1.7 and U-937 exposed to homochelidonine. Unexpectedly, homochelidonine and chelidonine-induced cytotoxicity was more pronounced in Jurkat cells contrary to MOLT-4 cells. Homochelidonine showed an antiproliferative effect on A549 cells but it was less effective compared to chelidonine. Biphasic dose-depended G1 and G2/M cell cycle arrest along with the population of sub-G1 was found after treatment with homochelidonine in MOLT-4 cells. In variance thereto, an increase in G2/M cells was detected after treatment with homochelidonine in Jurkat cells. Treatment with chelidonine induced cell cycle arrest in the G2/M cell cycle in both MOLT-4 and Jurkat cells. MOLT-4 and Jurkat cells treated with homochelidonine and chelidonine showed features of apoptosis such as phosphatidylserine exposure, a loss of mitochondrial membrane potential and an increase in the caspases -3/7, -8 and -9. Western blots indicate that homochelidonine and chelidonine exposure activates Chk1 and Chk2. Studies conducted with fluorescence microscopy demonstrated that chelidonine and homochelidonine inhibit tubulin polymerization in A549 cells. Collectively, the data indicate that chelidonine and homochelidonine are potent inducers of cell death in cancer cell lines, highlighting their potential relevance in leukemic cells.
Product Description
A549 Cell Line human, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 86012804, human lung (carcinoma), Epithelial
MOLT-4, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
RAJI Cell Line human, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 85011429, human lymph, Lymphoblastoid