- Carboxy-peptidase B. 4. Purification and characterization of the porcine enzyme.
Carboxy-peptidase B. 4. Purification and characterization of the porcine enzyme.
The Journal of biological chemistry (1960-08-01)
J E FOLK, K A PIEZ, W R CARROLL, J A GLADNER
PMID13823740