Merck
  • IER5 generates a novel hypo-phosphorylated active form of HSF1 and contributes to tumorigenesis.
IER5 generates a novel hypo-phosphorylated active form of HSF1 and contributes to tumorigenesis.
Scientific reports (2016-01-13)
Yoshinori Asano, Tatsuya Kawase, Atsushi Okabe, Shuichi Tsutsumi, Hitoshi Ichikawa, Satoko Tatebe, Issay Kitabayashi, Fumio Tashiro, Hideo Namiki, Tadashi Kondo, Kentaro Semba, Hiroyuki Aburatani, Yoichi Taya, Hitoshi Nakagama, Rieko Ohki
ABSTRACT
The transcription factors HSF1 and p53 both modulate the stress response, thereby protecting and facilitating the recovery of stressed cells, but both have the potential to promote tumor development. Here we show that a p53 target gene, IER5, encodes an activator of HSF1. IER5 forms a ternary complex with HSF1 and the phosphatase PP2A, and promotes the dephosphorylation of HSF1 at numbers of serine and threonine residues, generating a novel, hypo-phosphorylated active form of HSF1. IER5 is also transcriptionally upregulated in various cancers, although this upregulation is not always p53-dependent. The IER5 locus is associated with a so-called super enhancer, frequently associated with hyperactivated oncogenes in cancer cell lines. Enhanced expression of IER5 induces abnormal HSF1 activation in cancer cells and contributes to the proliferation of these cells under stressed conditions. These results reveal the existence of a novel IER5-mediated cancer regulation pathway that is responsible for the activation of HSF1 observed in various cancers.
Sigma-Aldrich
IGEPAL® CA-630, Molecular Biology
Sigma-Aldrich
Ethylene glycol-bis(succinic acid N-hydroxysuccinimide ester), powder
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
E-64d, protease inhibitor
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-PP2A Antibody, C subunit, clone 1D6, clone 1D6, Upstate®, from mouse