Merck
  • A dual phosphorylation switch controls 14-3-3-dependent cell surface expression of TASK-1.
A dual phosphorylation switch controls 14-3-3-dependent cell surface expression of TASK-1.
Journal of cell science (2016-01-09)
Markus Kilisch, Olga Lytovchenko, Eric C Arakel, Daniela Bertinetti, Blanche Schwappach
ABSTRACT
The transport of the K(+) channels TASK-1 and TASK-3 (also known as KCNK3 and KCNK9, respectively) to the cell surface is controlled by the binding of 14-3-3 proteins to a trafficking control region at the extreme C-terminus of the channels. The current model proposes that phosphorylation-dependent binding of 14-3-3 sterically masks a COPI-binding motif. However, the direct effects of phosphorylation on COPI binding and on the binding parameters of 14-3-3 isoforms are still unknown. We find that phosphorylation of the trafficking control region prevents COPI binding even in the absence of 14-3-3, and we present a quantitative analysis of the binding of all human 14-3-3 isoforms to the trafficking control regions of TASK-1 and TASK-3. Surprisingly, the affinities of 14-3-3 proteins for TASK-1 are two orders of magnitude lower than for TASK-3. Furthermore, we find that phosphorylation of a second serine residue in the C-terminus of TASK-1 inhibits 14-3-3 binding. Thus, phosphorylation of the trafficking control region can stimulate or inhibit transport of TASK-1 to the cell surface depending on the target serine residue. Our findings indicate that control of TASK-1 trafficking by COPI, kinases, phosphatases and 14-3-3 proteins is highly dynamic.
Sigma-Aldrich
(±)-2-(p-Methoxyphenoxy)propionic acid, ≥98%
Sigma-Aldrich
Monoclonal Anti-β-COP antibody produced in mouse, clone maD, ascites fluid