Merck
  • New uses for older antibiotics: nitrofurantoin, amikacin, colistin, polymyxin B, doxycycline, and minocycline revisited.
New uses for older antibiotics: nitrofurantoin, amikacin, colistin, polymyxin B, doxycycline, and minocycline revisited.
The Medical clinics of North America (2006-11-23)
Burke A Cunha
ABSTRACT
Nitrofurantoin, amikacin, colistin, polymyxin B, doxycycline, and minocycline are antibiotics with proven effectiveness against selected pathogens. These antibiotics have not developed resistance over time. As "low-resistance potential antibiotics" that are effective against an increasing number of infections due to resistant gram-positive or gram-negative pathogens, these antimicrobials remain an important part of the antibiotic armamentarium. They will be used increasingly in the future, as highly resistant organisms continue to be important clinically and therapeutic options remain limited.
Sigma-Aldrich
Nitrofurantoin, 98.0-102.0% (EP, UV), meets EP testing specifications
Sigma-Aldrich
Amikacin hydrate, aminoglycoside antibiotic
Supelco
Nitrofurantoin, VETRANAL®, analytical standard
USP
Nitrofurantoin, United States Pharmacopeia (USP) Reference Standard
Supelco
Nitrofurantoin, Pharmaceutical Secondary Standard; Certified Reference Material
Amikacin, European Pharmacopoeia (EP) Reference Standard