Merck
  • Lipoxygenase inhibitory constituents of the fruits of noni (Morinda citrifolia) collected in Tahiti.
Lipoxygenase inhibitory constituents of the fruits of noni (Morinda citrifolia) collected in Tahiti.
Journal of natural products (2007-03-24)
Shixin Deng, 'Afa K Palu, Brett J West, Chen X Su, Bing-Nan Zhou, Jarakae C Jensen
ABSTRACT
A phytochemical study of the fruits of noni (Morinda citrifolia) collected in Tahiti led to the isolation of two new lignans, (+)-3,4,3',4'-tetrahydroxy-9,7'alpha-epoxylignano-7 alpha,9'-lactone (1) and (+)-3,3'-bisdemethyltanegool (2), as well as seven known compounds, (-)-pinoresinol (3), (-)-3,3'-bisdemethylpinoresinol (4), quercetin (5), kaempferol (6), scopoletin (7), isoscopoletin (8), and vanillin. The structures of 1 and 2 were determined by spectroscopic techniques. Compounds 3, 6, and 8 were isolated for the first time from noni fruit. Compounds 1-8 were shown to inhibit 5- and/or 15-lipoxygenase, with IC50 values ranging from 0.43 to 16.5 microM. Compound 5 exhibited weak inhibitory activity toward cyclooxygenase-2.
Sigma-Aldrich
Nordihydroguaiaretic acid, ≥97.0% (HPLC)
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Scopoletin, ≥99%