Merck
  • Micron-scale plasma membrane curvature is recognized by the septin cytoskeleton.
Micron-scale plasma membrane curvature is recognized by the septin cytoskeleton.
The Journal of cell biology (2016-04-06)
Andrew A Bridges, Maximilian S Jentzsch, Patrick W Oakes, Patricia Occhipinti, Amy S Gladfelter
ABSTRACT
Cells change shape in response to diverse environmental and developmental conditions, creating topologies with micron-scale features. Although individual proteins can sense nanometer-scale membrane curvature, it is unclear if a cell could also use nanometer-scale components to sense micron-scale contours, such as the cytokinetic furrow and base of neuronal branches. Septins are filament-forming proteins that serve as signaling platforms and are frequently associated with areas of the plasma membrane where there is micron-scale curvature, including the cytokinetic furrow and the base of cell protrusions. We report here that fungal and human septins are able to distinguish between different degrees of micron-scale curvature in cells. By preparing supported lipid bilayers on beads of different curvature, we reconstitute and measure the intrinsic septin curvature preference. We conclude that micron-scale curvature recognition is a fundamental property of the septin cytoskeleton that provides the cell with a mechanism to know its local shape.
Product Description
Avanti
Egg Liss Rhod PE, Avanti Research - A Croda Brand
Avanti
Egg PC, Avanti Research - A Croda Brand
Avanti
Egg PC, Avanti Research - A Croda Brand
Avanti
Egg Liss Rhod PE, Avanti Research - A Croda Brand, powder
Avanti
Liver PI, Avanti Research - A Croda Brand
Avanti
Liver PI, Avanti Research - A Croda Brand
Avanti
18:1 DGS-NTA(Ni), Avanti Research - A Croda Brand
Avanti
18:1 DGS-NTA(Ni), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand