Merck
  • Iridium-catalyzed synthesis of primary allylic amines from allylic alcohols: sulfamic acid as ammonia equivalent.
Iridium-catalyzed synthesis of primary allylic amines from allylic alcohols: sulfamic acid as ammonia equivalent.
Angewandte Chemie (International ed. in English) (2007-03-14)
Christian Defieber, Martin A Ariger, Patricia Moriel, Erick M Carreira
Sigma-Aldrich
(S)-(−)-1,1′-Bi(2-naphthol), 99%
Sigma-Aldrich
Sulfamic acid, ACS reagent, 99.3%
Sigma-Aldrich
Sulfamic acid, reagent grade, 98%
Sigma-Aldrich
Sulfamic acid, ReagentPlus®, ≥99%