Merck
  • A high-throughput photodynamic therapy screening platform with on-chip control of multiple microenvironmental factors.
A high-throughput photodynamic therapy screening platform with on-chip control of multiple microenvironmental factors.
Lab on a chip
Lou X, et al.