Merck
  • Pluripotency markers are differentially induced by MEK inhibition in thyroid and melanoma BRAFV600E cell lines.
Pluripotency markers are differentially induced by MEK inhibition in thyroid and melanoma BRAFV600E cell lines.
Cancer biology & therapy (2016-02-02)
Emma R Dorris, Gordon Blackshields, Gary Sommerville, Mohsen Alhashemi, Andrew Dias, Victoria McEneaney, Paul Smyth, John J O'Leary, Orla Sheils
ABSTRACT
Oncogenic mutations in BRAF are common in melanoma and thyroid carcinoma and drive constitutive activation of the MAPK pathway. Molecularly targeted therapies of this pathway improves survival compared to chemotherapy; however, responses tend to be short-lived as resistance invariably occursCell line models of melanoma and thyroid carcinoma, +/- BRAF(V600E) activating mutation, were treated with the MEK inhibitor PD0325901. Treated and naive samples were assayed for expression of key members of the MAPK pathway. Global microRNA expression profiling of naive and resistant cells was performed via next generation sequencingand indicated pluripotency pathways in resistance. Parental cell lines were progressed to holoclones to confirm the miRNA stemness profileMembers of the MIR302/373/374/520 family of embryonic stem cell specific cell cycle regulating (ESCC) microRNAs were identified as differentially expressed between resistant BRAF(V600E) melanoma and thyroid cell lines. Upregulated expression of gene and protein stemness markers, upregulated expression of MAPK pathway genes and downregulation of the ESCC MIR302 cluster in BRAF(V600E) melanoma indicated an increased stem-like phenotype in resistant BRAF(V600E) melanoma. Conversely, downregulated expression of gene and protein stemness markers, downregulated expression of MAPK pathway genes, upregulation of the ESCC MIR520 cluster, reeexpression of cell surface receptors, and induced differentiation-associated morphology in resistant BRAF(V600E) indicate a differentiated phenotype associated with MEK inhibitor resistance in BRAF(V600E) thyroid cellsThe differential patterns of resistance observed between BRAF(V600E) melanoma and thyroid cell lines may reflect tissue type or de novo differentiation, but could have significant impact on the response of primary and metastatic cells to MEK inhibitor treatment. This study provides a basis for the investigation of the cellular differentiation/self-renewal access and its role in resistance to MEK inhibition.
Sigma-Aldrich
Anti-ALDH1A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution