Merck
  • Evaluating heavy metal contents in nine composts using four digestion methods.
Evaluating heavy metal contents in nine composts using four digestion methods.
Bioresource technology (2004-06-23)
Zeng-Yei Hseu
ABSTRACT
The production and application of compost potentially contaminate the environment with heavy metals. The heavy metal contents of composts must be exactly determined. Therefore, this study aims to understand the heavy metal contents in composts, using different digestion methods and to recommend the most appropriate digestion method making this measurement. Nine composts from different sources, including swine manure, poultry litter, food waste, municipal sewage sludge, soybean meal, wood residues, flesh-bone dust, fish-bone meal and guano, were selected to be digested by four methods to determine the contents of Cd, Cr, Cu, Mn, Ni, Pb and Zn by AAS. The four digestion methods were nitric acid, dry ashing, nitric-perchloric acid and sulfuric acid methods. Analytical results indicated that the nitric acid procedure was the most efficient for recovering Cd, Mn and Ni from most compost samples. The sulfuric acid procedure yielded the lowest recovery of Pb from the certified reference material (BCR 146) and the compost samples, especially from the composts with high pH and EC values potentially high in Ca. The nitric acid procedure was recommended as the method for digesting the composts herein, based on recovery analysis, cost and time taken. Dry ashing was recommended as a flexible method. Nitric-perchloric acid procedure was not recommended because perchloric acid is potentially hazardous during digestion and it recovers relatively little heavy metal.