Merck
  • E2F1 in neurons is cleaved by calpain in an NMDA receptor-dependent manner in a model of HIV-induced neurotoxicity.
E2F1 in neurons is cleaved by calpain in an NMDA receptor-dependent manner in a model of HIV-induced neurotoxicity.
Journal of neurochemistry (2014-10-04)
Jacob W Zyskind, Ying Wang, Giyong Cho, Jenhao H Ting, Dennis L Kolson, David R Lynch, Kelly L Jordan-Sciutto
ABSTRACT
The transcription factor E2F1 activates gene targets required for G1 -S phase progression and for apoptosis, and exhibits increased expression levels in neurons in several CNS diseases including HIV encephalitis, Alzheimer disease, and Parkinson's Disease. While E2F1 is known to regulate cell viability through activation of caspases, here we present evidence supporting the involvement of E2F1 in N-methyl-d-aspartate (NMDA) receptor-dependent, HIV-induced neuronal death mediated by calpains. Using an in vitro model of HIV-induced neurotoxicity that is dependent on NMDA receptor and calpain activation, we have shown that cortical neurons lacking functional E2F1 are less susceptible to neuronal death. In addition, we report that neuronal E2F1 is cleaved by calpain to a stable 55-kiloDalton fragment following NR2B-dependent NMDA receptor stimulation. This cleavage of E2F1 is protein conformation-dependent and involves at least two cleavage events, one at each terminus of the protein. Intriguingly, the stabilized E2F1 cleavage product is produced in post-mitotic neurons of all ages, but fails to be stabilized in cycling cells. Finally, we show that a matching E2F1 cleavage product is produced in human fetal neurons, suggesting that calpain cleavage of E2F1 may be produced in human cortical tissue. These results suggest neuronal E2F1 is processed in a novel manner in response to NMDA receptor-mediated toxicity, a mechanism implicated in HIV-associated neurocognitive disorders pathogenesis as well as several other diseases of the CNS. After crossing the blood-brain barrier, HIV-infected monocytes differentiate into macrophages and release excitotoxins and inflammatory factors including glutamate into the brain parenchyma (1). These factors stimulate neuronal N-Methyl-d-aspartate (NMDA) receptors (2), causing calcium influx (3) and subsequent activation of the cysteine protease calpain (4). Activated calpain cleaves multiple substrates including E2F1, producing a stabilized protein fragment with truncations at the N- and C-terminus (5). Calpain-cleaved E2F1 may contribute to calpain-mediated neuronal damage observed in NMDA receptor-mediated neurotoxicity (6).
Sigma-Aldrich
Monoclonal Anti-MAT-Tag® antibody produced in mouse, purified immunoglobulin, ~2 mg/mL, clone MAT1-87
Sigma-Aldrich
Calpain-1, Porcine Erythrocytes, Calpain-1, Porcine Erythrocytes, is a native calpain-1. A heterodimeric cysteine proteinase with low Ca2+ requirement (EC50 = 2 µM).
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution