Merck
  • Niacin-mediated Tace activation ameliorates CMT neuropathies with focal hypermyelination.
Niacin-mediated Tace activation ameliorates CMT neuropathies with focal hypermyelination.
EMBO molecular medicine (2016-11-02)
Alessandra Bolino, Françoise Piguet, Valeria Alberizzi, Marta Pellegatta, Cristina Rivellini, Marta Guerrero-Valero, Roberta Noseda, Chiara Brombin, Alessandro Nonis, Patrizia D'Adamo, Carla Taveggia, Stefano Carlo Previtali
ABSTRACT
Charcot-Marie-Tooth (CMT) neuropathies are highly heterogeneous disorders caused by mutations in more than 70 genes, with no available treatment. Thus, it is difficult to envisage a single suitable treatment for all pathogenetic mechanisms. Axonal Neuregulin 1 (Nrg1) type III drives Schwann cell myelination and determines myelin thickness by ErbB2/B3-PI3K-Akt signaling pathway activation. Nrg1 type III is inhibited by the α-secretase Tace, which negatively regulates PNS myelination. We hypothesized that modulation of Nrg1 levels and/or secretase activity may constitute a unifying treatment strategy for CMT neuropathies with focal hypermyelination as it could restore normal levels of myelination. Here we show that in vivo delivery of Niaspan, a FDA-approved drug known to enhance TACE activity, efficiently rescues myelination in the Mtmr2-/- mouse, a model of CMT4B1 with myelin outfoldings, and in the Pmp22+/- mouse, which reproduces HNPP (hereditary neuropathy with liability to pressure palsies) with tomacula. Importantly, we also found that Niaspan reduces hypermyelination of Vim (vimentin)-/- mice, characterized by increased Nrg1 type III and Akt activation, thus corroborating the hypothesis that Niaspan treatment downregulates Nrg1 type III signaling.
Sigma-Aldrich
MISSION® esiRNA, targeting human NRG1
Sigma-Aldrich
Anti-Calnexin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 82-87, culture supernatant, clone 12, Chemicon®
Sigma-Aldrich
Anti-Po Antibody, Chemicon®, from chicken
Sigma-Aldrich
Anti-Vinculin Antibody, clone V284, clone V284, Upstate®, from mouse