Merck
  • Potential biological role of transforming growth factor-beta1 in human congenital kidney malformations.
Potential biological role of transforming growth factor-beta1 in human congenital kidney malformations.
The American journal of pathology (2000-11-14)
S P Yang, A S Woolf, H T Yuan, R J Scott, R A Risdon, M J O'Hare, P J Winyard
ABSTRACT
Transformations between epithelial and mesenchymal cells are widespread during normal development and adult disease, and transforming growth factor-beta1 (TGF-beta1) has been implicated in some of these phenotypic switches. Dysplastic kidneys are a common cause of chronic kidney failure in young children and result from perturbed epithelial-mesenchymal interactions. In this study, we found that components of the TGF-beta1 axis were expressed in these malformations: TGF-beta1 mRNA and protein were up-regulated in dysplastic epithelia and surrounding mesenchymal cells, whereas TGF-beta receptors I and II were expressed in aberrant epithelia. We generated a dysplastic kidney epithelial-like cell line that expressed cytokeratin, ZO1, and MET, and found that exogenous TGF-beta1 inhibited proliferation and decreased expression of PAX2 and BCL2, molecules characterizing dysplastic tubules in vivo. Furthermore, addition of TGF-beta1 specifically induced morphological changes compatible with a shift to a mesenchymal phenotype, accompanied by loss of ZO1 at cell borders and up-regulation of the mesenchymal markers alpha-smooth muscle actin and fibronectin. The descriptive and functional data presented in this report potentially implicate TGF-beta1 in the pathobiology of dysplastic kidneys and our results provide preliminary evidence that an epithelial-to-mesenchymal phenotypic switch may be implicated in a clinically important developmental aberration.
Sigma-Aldrich
Monoclonal Anti-Fibronectin antibody produced in mouse, clone IST-3, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Cytokeratin, pan (Mixture) antibody produced in mouse, clone C-11+PCK-26+CY-90+KS-1A3+M20+A53-B/A2, ascites fluid
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Alkaline Phosphatase antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid