Merck
  • Forkhead Box C1 Regulates Human Primary Keratinocyte Terminal Differentiation.
Forkhead Box C1 Regulates Human Primary Keratinocyte Terminal Differentiation.
PloS one (2016-12-03)
Lianghua Bin, Liehua Deng, Hengwen Yang, Leqing Zhu, Xiao Wang, Michael G Edwards, Brittany Richers, Donald Y M Leung
ABSTRACT
The epidermis serves as a critical protective barrier between the internal and external environment of the human body. Its remarkable barrier function is established through the keratinocyte (KC) terminal differentiation program. The transcription factors specifically regulating terminal differentiation remain largely unknown. Using a RNA-sequencing (RNA-seq) profiling approach, we found that forkhead box c 1 (FOXC1) was significantly up-regulated in human normal primary KC during the course of differentiation. This observation was validated in human normal primary KC from several different donors and human skin biopsies. Silencing FOXC1 in human normal primary KC undergoing differentiation led to significant down-regulation of late terminal differentiation genes markers including epidermal differentiation complex genes, keratinization genes, sphingolipid/ceramide metabolic process genes and epidermal specific cell-cell adhesion genes. We further demonstrated that FOXC1 works down-stream of ZNF750 and KLF4, and upstream of GRHL3. Thus, this study defines FOXC1 as a regulator specific for KC terminal differentiation and establishes its potential position in the genetic regulatory network.
Sigma-Aldrich
MISSION® esiRNA, targeting human ZNF750
Sigma-Aldrich
MISSION® esiRNA, targeting human KLF4