Merck
  • Tumour-processed osteopontin and lactadherin drive the protumorigenic reprogramming of microglia and glioma progression.
Tumour-processed osteopontin and lactadherin drive the protumorigenic reprogramming of microglia and glioma progression.
Oncogene (2016-04-05)
A Ellert-Miklaszewska, P Wisniewski, M Kijewska, P Gajdanowicz, D Pszczolkowska, P Przanowski, M Dabrowski, M Maleszewska, B Kaminska
ABSTRACT
Tumour tissue is infiltrated by myeloid cells that are reprogrammed into alternatively activated/regenerative (M2) macrophages. The contribution of major signalling pathways and their modulators/targets involved in the macrophage reprogramming is poorly known. Glioblastoma (malignant brain tumour) attracts and reprograms brain-resident microglia and peripheral macrophages into cells that increase invasion, angiogenesis and suppress antitumour immunity. Using a 'function-first' approach and glioma secretome proteomics we identified osteopontin and lactadherin as proteins that cooperatively activate amoeboid transformation, phagocytosis and motility of primary microglia cultures via integrins and FAK-Akt (focal adhesion kinase-Akt) signalling. A synthetic peptide interfering with integrin ligands blocks glioma-microglia communication, functional activation and M2 gene expression. We found that osteopontin/secreted phosphoprotein 1 (Spp1) produced by non-transformed cells acts as a proinflammatory factor inducing inflammatory signalling and M1 genes, and counteracts the action of lactadherin. Using constructs encoding functional mutants of osteopontin, we demonstrated sequential processing of Spp1 by thrombin and matrix metalloproteinase-3 and/or -7 (MMP-3 and/or -7) in glioma cells, which generates a microglia-activating form devoid of the inflammatory activity, while retaining the M2 reprogramming potential. A similar form of osteopontin is secreted by human glioma cells but not normal human astrocytes. Knockdown of osteopontin or lactadherin in glioma cells reduces intracranial glioma growth, blocks amoeboid transformation of myeloid cells and affects M2 reprogramming of microglia/macrophages. Our findings demonstrate how glioma cells misuse macrophage-activating signals and redesign primarily proinflammatory signals towards their advantage to induce M2 reprogramming of tumour-infiltrating brain macrophages.
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Toluidine Blue, suitable for microscopy, 8.74% (ZN (THEORY))
Sigma-Aldrich
MISSION® esiRNA, targeting human MFGE8