Merck
  • Transforming Growth Factor (TGF)-β Promotes de Novo Serine Synthesis for Collagen Production.
Transforming Growth Factor (TGF)-β Promotes de Novo Serine Synthesis for Collagen Production.
The Journal of biological chemistry (2016-11-12)
Recep Nigdelioglu, Robert B Hamanaka, Angelo Y Meliton, Erin O'Leary, Leah J Witt, Takugo Cho, Kaitlyn Sun, Catherine Bonham, David Wu, Parker S Woods, Aliya N Husain, Don Wolfgeher, Nickolai O Dulin, Navdeep S Chandel, Gökhan M Mutlu
ABSTRACT
TGF-β promotes excessive collagen deposition in fibrotic diseases such as idiopathic pulmonary fibrosis (IPF). The amino acid composition of collagen is unique due to its high (33%) glycine content. Here, we report that TGF-β induces expression of glycolytic genes and increases glycolytic flux. TGF-β also induces the expression of the enzymes of the de novo serine synthesis pathway (phosphoglycerate dehydrogenase (PHGDH), phosphoserine aminotransferase 1 (PSAT1), and phosphoserine phosphatase (PSPH)) and de novo glycine synthesis (serine hydroxymethyltransferase 2 (SHMT2)). Studies in fibroblasts with genetic attenuation of PHGDH or SHMT2 and pharmacologic inhibition of PHGDH showed that these enzymes are required for collagen synthesis. Furthermore, metabolic labeling experiments demonstrated carbon from glucose incorporated into collagen. Lungs from humans with IPF demonstrated increased expression of PHGDH and SHMT2. These results indicate that the de novo serine synthesis pathway is necessary for TGF-β-induced collagen production and suggest that this pathway may be a therapeutic target for treatment of fibrotic diseases including IPF.
Sigma-Aldrich
MISSION® esiRNA, targeting human PHGDH
Sigma-Aldrich
SB 431542 hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Goat Anti-Rabbit IgG, H & L Chain Specific Peroxidase Conjugate, liquid, Calbiochem®
Sigma-Aldrich
Sodium oxamate, ≥98%
Sigma-Aldrich
MISSION® esiRNA, targeting human LDHA
Sigma-Aldrich
MISSION® esiRNA, targeting human SHMT1
Sigma-Aldrich
MISSION® esiRNA, targeting human SHMT2
Sigma-Aldrich
MISSION® esiRNA, targeting human SMAD3
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Goat Anti-Mouse IgG, H&L Chain Specific Peroxidase Conjugate, liquid, Calbiochem®
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid