Merck
  • Dietary intake of phosphorus flame retardants (PFRs) using Swedish food market basket estimations.
Dietary intake of phosphorus flame retardants (PFRs) using Swedish food market basket estimations.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2016-12-15)
Giulia Poma, Anders Glynn, Govindan Malarvannan, Adrian Covaci, Per Ola Darnerud
ABSTRACT
The occurrence of eight phosphorus flame retardants (PFRs) was investigated in 53 composite food samples from 12 food categories, collected in 2015 for a Swedish food market basket study. 2-ethylhexyl diphenyl phosphate (EHDPHP), detected in most food categories, had the highest median concentrations (9 ng/g ww, pastries). It was followed by triphenyl phosphate (TPHP) (2.6 ng/g ww, fats/oils), tris(1,3-dichloro-2-propyl) phosphate (TDCIPP) (1.0 ng/g ww, fats/oils), tris(2-chloroethyl) phosphate (TCEP) (1.0 ng/g ww, fats/oils), and tris(1-chloro-2-propyl) phosphate (TCIPP) (0.80 ng/g ww, pastries). Tris(2-ethylhexyl) phosphate (TEHP), tri-n-butyl phosphate (TNBP), and tris(2-butoxyethyl) phosphate (TBOEP) were not detected in the analyzed food samples. The major contributor to the total dietary intake was EHDPHP (57%), and the food categories which contributed the most to the total intake of PFRs were processed food, such as cereals (26%), pastries (10%), sugar/sweets (11%), and beverages (17%). The daily per capita intake of PFRs (TCEP, TPHP, EHDPHP, TDCIPP, TCIPP) from food ranged from 406 to 3266 ng/day (or 6-49 ng/kg bw/day), lower than the health-based reference doses. This is the first study reporting PFR intakes from other food categories than fish (here accounting for 3%). Our results suggest that the estimated human dietary exposure to PFRs may be equally important to the ingestion of dust.
Sigma-Aldrich
2-Methylheptane, 98%