Merck
  • Bronchial inflammation in acute bacterial exacerbations of chronic bronchitis: the role of leukotriene B4.
Bronchial inflammation in acute bacterial exacerbations of chronic bronchitis: the role of leukotriene B4.
The European Respiratory Journal (2000)
Crooks SW