Merck
  • Characterization and expression pattern of the frizzled gene Fzd9, the mouse homolog of FZD9 which is deleted in Williams-Beuren syndrome.
Characterization and expression pattern of the frizzled gene Fzd9, the mouse homolog of FZD9 which is deleted in Williams-Beuren syndrome.
Genomics (1999-04-13)
Y K Wang, R Spörle, T Paperna, K Schughart, U Francke
ABSTRACT
The frizzled gene family is conserved from insects to mammals and codes for putative Wnt receptors that share a cysteine-rich extracellular domain and seven transmembrane domains. We previously identified a novel frizzled gene, FZD3, now renamed FZD9, in the Williams-Beuren syndrome (WBS) deletion region at chromosomal band 7q11.23 and showed that its product can interact with the Drosophila wingless protein. Here, we report the characterization of the mouse homolog Fzd9. The Fzd9 gene produces a 2.4-kb transcript encoding a 592-amino-acid protein with 95% identity to the human FZD9. Fzd9 was mapped to the conserved syntenic region on distal mouse chromosome 5. By RNA in situ hybridization studies of whole-mount embryos and sections we delineated the temporal and spatial expression patterns in the neural tube, trunk skeletal muscle precursors (myotomes), limb skeletal anlagen, craniofacial regions, and nephric ducts. In adult mouse tissue, the Fzd9 transcript is abundantly present in heart, brain, testis, and skeletal muscle. In testis, Fzd9 is expressed in all spermatogenic cell types. Immunohistochemical studies of cells transfected with a Fzd9 expression construct confirm that Fzd9 is a membrane protein. These results suggest potential Wnt ligands of Fzd9, a role of Fzd9 in skeletal muscle specification, and contributions of FZD9 to the WBS phenotype.