Merck
  • MiR133b is involved in endogenous hydrogen sulfide suppression of sFlt-1 production in human placenta.
MiR133b is involved in endogenous hydrogen sulfide suppression of sFlt-1 production in human placenta.
Placenta (2017-04-30)
Tian-Xiao Hu, Xuejing Guo, Gang Wang, Lu Gao, Ping He, Yang Xia, Hang Gu, Xin Ni
ABSTRACT
Increased production of soluble fms-like tyrosine kinase-1 (sFlt-1) from placenta is one of the major contributors to the development of preeclampsia. Our previous study has shown that hydrogen sulfide (H
Sigma-Aldrich
5,6-Dichlorobenzimidazole 1-β-D-ribofuranoside
Sigma-Aldrich
MISSION® esiRNA, targeting human CTH
Sigma-Aldrich
MISSION® esiRNA, targeting human CBS