Merck
  • A study to evaluate the potential of an in silico approach for predicting dipeptidyl peptidase-IV inhibitory activity in vitro of protein hydrolysates.
A study to evaluate the potential of an in silico approach for predicting dipeptidyl peptidase-IV inhibitory activity in vitro of protein hydrolysates.
Food chemistry (2017-05-30)
Tzu-Yuan Wang, Cheng-Hong Hsieh, Chuan-Chuan Hung, Chia-Ling Jao, Pei-Yi Lin, You-Liang Hsieh, Kuo-Chiang Hsu
ABSTRACT
A total of 294 edible protein sequences and 5 commercial proteases listed in the BIOPEP database were analyzed in silico. The frequency (A), a parameter in silico described previously, was examined further to calculating the ratio of truncated peptides with Xaa-proline and/or Xaa-alanine to all peptide fragments in a protein hydrolyzed with a protease, using the BIOPEP database. Then the in vitro DPP-IV inhibitory activity was determined using the same 15 protein and protease combinations to evaluate their relationship. The result shows that A values considering the number of Xaa-proline+Xaa-alanine exhibited a strong correlation with in vitro DPP-IV inhibition rates by Pearson's correlation analysis (r=0.6993; P<0.05). Therefore, the in silico approach is effective to predict DPP-IV inhibitory activities in vitro of protein hydrolysates.
Sigma-Aldrich
Papain from papaya latex, crude powder, 1.5-10 units/mg solid
Sigma-Aldrich
Nα-Benzoyl-L-arginine ethyl ester hydrochloride, trypsin substrate
Sigma-Aldrich
κ-Casein from bovine milk, ≥70% (PAGE), lyophilized powder
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Thermolysin from Geobacillus stearothermophilus, Type X, lyophilized powder, 30-350 units/mg protein (E1%/280)
Sigma-Aldrich
α-Casein from bovine milk, ≥70% αs-casein basis (electrophoresis), lyophilized powder
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
β-Casein from bovine milk, BioUltra, ≥98% (PAGE)
Sigma-Aldrich
Myoglobin from equine skeletal muscle, 95-100%, essentially salt-free, lyophilized powder
Sigma-Aldrich
Bromelain from pineapple stem, ≥3 units/mg protein
Sigma-Aldrich
β-Lactoglobulin from bovine milk, ≥90% (PAGE), lyophilized powder