Merck
Exopolysaccharide from
Journal of immunology (Baltimore, Md. : 1950) (2017-02-17)
Mallory L Paynich, Sara E Jones-Burrage, Katherine L Knight
ABSTRACT
Commensal bacteria contribute to immune homeostasis in the gastrointestinal tract; however, the underlying mechanisms for this are not well understood. A single dose of exopolysaccharide (EPS) from the probiotic spore-forming bacterium