Merck
  • Deficiency of pigment epithelium-derived factor in nasopharyngeal carcinoma cells triggers the epithelial-mesenchymal transition and metastasis.
Deficiency of pigment epithelium-derived factor in nasopharyngeal carcinoma cells triggers the epithelial-mesenchymal transition and metastasis.
Cell death & disease (2017-06-02)
Ting Zhang, Ping Yin, Zichen Zhang, Banglao Xu, Di Che, Zhiyu Dai, Chang Dong, Ping Jiang, Honghai Hong, Zhonghan Yang, Ti Zhou, Jianyong Shao, Zumin Xu, Xia Yang, Guoquan Gao
ABSTRACT
Distant metastasis is the primary cause of nasopharyngeal carcinoma (NPC) treatment failure while epithelial-mesenchymal transition (EMT) is the critical process of NPC invasion and metastasis. However, tumor-suppressor genes involved in the EMT and metastasis of NPC have not been explored clearly compared with the oncogenes. In the present study, the expression of pigment epithelium-derived factor (PEDF), a potent endogenous antitumor factor, was diminished in human NPC tissues and associated with clinicopathological and EMT features. The knockdown of PEDF induced EMT in lower metastatic NPC cell lines and overexpression of PEDF restored epithelial phenotype in higher metastatic NPC cell lines with typical EMT. The inhibition of PEDF mediated NPC cell spontaneous metastasis in vivo. LRP6/GSK3β/β-catenin signal pathway rather than AKT/GSK3β pathway was involved in the effects of PEDF on EMT. The expression of PEDF was directly downregulated by elevated miR-320c in NPC. In conclusion, our findings indicate for the first time that PEDF functions as tumor-suppressor gene in the occurrence of EMT and metastasis in NPC. PEDF could serve as a promising candidate for NPC diagnosis, prognosis and treatment.
Sigma-Aldrich
MISSION® esiRNA, targeting human SERPINF1
Sigma-Aldrich
2-Phenylindole, technical grade, 95%