- Offspring from sequential matings between Bacillus thuringiensis-resistant and Bacillus thuringiensis-susceptible Heliothis virescens moths (Lepidoptera: Noctuidae).
Offspring from sequential matings between Bacillus thuringiensis-resistant and Bacillus thuringiensis-susceptible Heliothis virescens moths (Lepidoptera: Noctuidae).
Journal of Economic Entomology (2010)
Blanco CA