Merck
  • Membrane permeabilization by trypanosome lytic factor, a cytolytic human high density lipoprotein.
Membrane permeabilization by trypanosome lytic factor, a cytolytic human high density lipoprotein.
The Journal of biological chemistry (2009-03-28)
John M Harrington, Sawyer Howell, Stephen L Hajduk
ABSTRACT
Trypanosome lytic factor (TLF) is a subclass of human high density lipoprotein (HDL) that mediates an innate immune killing of certain mammalian trypanosomes, most notably Trypanosoma brucei brucei, the causative agent of a wasting disease in cattle. Mechanistically, killing is initiated in the lysosome of the target trypanosome where the acidic pH facilitates a membrane-disrupting activity by TLF. Here we utilize a model liposome system to characterize the membrane binding and permeabilizing activity of TLF and its protein constituents, haptoglobin-related protein (Hpr), apolipoprotein L-1 (apoL-1), and apolipoprotein A-1 (apoA-1). We show that TLF efficiently binds and permeabilizes unilamellar liposomes at lysosomal pH, whereas non-lytic human HDL exhibits inefficient permeabilizing activity. Purified, delipidated Hpr and apoL-1 both efficiently permeabilize lipid bilayers at low pH. Trypanosome lytic factor, apoL-1, and apoA-1 exhibit specificity for anionic membranes, whereas Hpr permeabilizes both anionic and zwitterionic membranes. Analysis of the relative particle sizes of susceptible liposomes reveals distinctly different membrane-active behavior for native TLF and the delipidated protein components. We propose that lysosomal membrane damage in TLF-susceptible trypanosomes is initiated by the stable association of the TLF particle with the lysosomal membrane and that this is a property unique to this subclass of human HDL.
Product Description
Avanti
Egg Trans PE, L-α-phosphatidylethanolamine, transphosphatidylated (Egg, Chicken), powder
Avanti
Egg PC, Avanti Research - A Croda Brand
Avanti
Egg Trans PE, L-α-phosphatidylethanolamine, transphosphatidylated (Egg, Chicken), chloroform
Avanti
Egg PC, Avanti Research - A Croda Brand
Avanti
Soy PC (20%), Avanti Research - A Croda Brand
Sigma-Aldrich
Haptoglobin Human, Phenotype 1-1, 98-100%, essentially salt-free, lyophilized powder
Avanti
Heart CA, Avanti Research - A Croda Brand 840012P, powder
Avanti
Egg PA, Avanti Research - A Croda Brand
Avanti
Egg PA, Avanti Research - A Croda Brand
Avanti
Brain PS, Avanti Research - A Croda Brand
Avanti
Brain PS, Avanti Research - A Croda Brand
Avanti
Heart CA, Avanti Research - A Croda Brand