Merck
  • Effect of substituted hydroxyl groups in the changes of solution turbidity in the oxidation of aromatic contaminants.
Effect of substituted hydroxyl groups in the changes of solution turbidity in the oxidation of aromatic contaminants.
Environmental science and pollution research international (2016-07-29)
N Villota, Lomas Jm, Camarero Lm
ABSTRACT
This paper deals with the changes of turbidity that are generated in aqueous solutions of phenol when they are oxidized by using different Fenton technologies. Results revealed that if the Fenton reaction was promoted with UV light, the turbidity that was generated in the water doubled. Alternatively, the use of ultrasonic waves produced an increase in turbidity which initially proceeded slowly, reaching intensities eight times higher than in the conventional Fenton treatment. As well, the turbidity showed a high dependence on pH. It is therefore essential to control acidity throughout the reaction. The maximum turbidity was generated when operating at pH = 2.0, and it slowly decreased with increasing to a value of pH = 3.0, at which the turbidity was the lowest. This result was a consequence of the presence of ferric ions in solution. At pH values greater than 3.5, the turbidity increased almost linearly until at pH = 5.0 reached its maximum intensity. In this range, ferrous ions may generate an additional contribution of radicals that promote the degradation of the phenol species that produce turbidity. Turbidity was enhanced at ratios R = 4.0 mol H
Sigma-Aldrich
2,5-Dihydroxy-1,4-benzoquinone, 98%
Sigma-Aldrich
1,2,4-Benzenetriol, ReagentPlus®, 99%