Merck
  • Enzyme-functionalized thin-cladding long-period fiber grating in transition mode at dispersion turning point for sugar-level and glucose detection.
Enzyme-functionalized thin-cladding long-period fiber grating in transition mode at dispersion turning point for sugar-level and glucose detection.
Journal of biomedical optics (2017-02-09)
Abdulyezir A Badmos, Qizhen Sun, Zhongyuan Sun, Junxi Zhang, Zhijun Yan, Petro Lutsyk, Alex Rozhin, Lin Zhang
ABSTRACT
Enzyme-functionalized dual-peak long-period fiber grating (LPFG) inscribed in 80 - ? m -cladding B/Ge codoped single-mode fiber is presented for sugar-level and specific glucose detection. Before enzyme functionalization, the dual-peak LPFG was employed for refractive index sensing and sugar-level detection and high sensitivities of ? 4298.20 ?? nm / RIU and 4.6696 ?? nm / % were obtained, respectively. Glucose detection probe was attained by surface functionalization of the dual-peak LPFG via covalent binding with aminopropyl triethoxysilane used as a binding site. Optical micrographs confirmed the presence of enzyme. The surface-functionalized dual-peak LPFG was tested with D-(+)-glucose solution of different concentrations. While the peak 2 at the longer wavelength was suitable only to measure lower glucose concentration (0.1 to 1.6 ?? mg / ml ) recording a high sensitivity of 12.21 ± 0.19 ?? nm / ( mg / ml ) , the peak 1 at the shorter wavelength was able to measure a wider range of glucose concentrations (0.1 to 3.2 ?? mg / ml ) exhibiting a maximum resonance wavelength shift of 7.12 ± 0.12 ?? nm / mg / ml . The enzyme-functionalized dual-peak LPFG has the advantage of direct inscription of highly sensitive grating structures in thin-cladding fibre without etching, and most significantly, its sensitivity improvement of approximately one order of magnitude higher than previously reported LPFG and excessively tilted fibre grating (Ex-TFG) for glucose detection.
Sigma-Aldrich
Sodium acetate buffer solution, pH 5.2±0.1 (25 °C), Molecular Biology, 3 M, 0.2 μm filtered
Sigma-Aldrich
Triethoxysilane, 95%