Merck
RNAi-mediated knockdown of
Oncotarget (2017-10-21)
Guang-Yu Bai, Si-Hang Song, Rui-Zhen Sun, Zi-Hui Zhang, Jingyu Li, Zhen-Dong Wang, Zhong-Hua Liu, Lei Lei
ABSTRACT
Somatic cell nuclear transfer is an important technique for life science research, but its efficiency is still extremely low, and most genes that are important during early development, such as X chromosome-linked genes, are not appropriately expressed during this process. Poly (ADP-ribose) polymerase (PARP) is an enzyme that transfers ADP ribose clusters to target proteins. PARP family members such as PARP1 participate in cellular signalling pathways through poly (ADP-ribosylation) (PARylation), which ultimately promotes changes in chromatin structure, gene expression, and the localization and activity of proteins that mediate signalling responses. PARP1 is associated with X chromosome inactivation (Xi). Here, we showed that abnormal Xi occurs in somatic cell nuclear transfer (NT) blastocysts, whereas in female blastocysts derived from cumulus cell nuclear transfer, both X chromosomes were inactive.
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Polyvinylpyrrolidone, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
Trypsin from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
Sigma-Aldrich
Mineral oil, light oil, (neat)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid
Sigma-Aldrich
Cytochalasin B from Drechslera dematioidea, ≥98% (HPLC), powder