Merck
  • Thermal experience during embryogenesis contributes to the induction of dwarfism in whitefish Coregonus lavaretus.
Thermal experience during embryogenesis contributes to the induction of dwarfism in whitefish Coregonus lavaretus.
PloS one (2017-09-26)
Peter Steinbacher, Josef Wanzenböck, Magdalena Brandauer, Raphael Holper, Jasmin Landertshammer, Magdalena Mayr, Christian Platzl, Walter Stoiber
ABSTRACT
Ecotype pairs provide well-suited model systems for study of intraspecific phenotypical diversification of animals. However, little is still known about the processes that account for the development of different forms and sizes within a species, particularly in teleosts. Here, embryos of a normal-growing 'large' form and a dwarf form of whitefish Coregonus lavaretus were incubated at two temperatures that are usually experienced at their own spawning sites (2°C for the normal and 6°C for the dwarf form). All fish were subjected to similar thermal treatment after hatching. The present data demonstrate for the first time that different thermal experience in embryonic life has lasting effects on body and muscle growth of this ecotype pair and contributes to the development of the dwarf form. Thus, juvenile fish of the regular form are much smaller and have less muscle mass when pre-hatching thermal conditions were similar to those typical for the spawning sites of the dwarf form (6°C) than when subjected to conditions of their own spawning sites (2°C). Surprisingly, fish of the dwarf form exhibit a similar pattern of response to thermal history (2°-fish much larger than 6°-fish), indicating that in their case, normal spawning site temperature (6°C) is indeed likely to act as a growth limiting factor. Results also demonstrate that the hypertrophic and hyperplastic muscle growth modes are similarly affected by thermal history. Immunolabelling experiments for Pax7, H3P and Mef2 provide evidence that the cellular mechanisms behind the increased growth rates after cold incubation in both ecotypes are increased proliferation and reduced differentiation rates of muscle precursor cells. This is of major significance to aspects of ecological and developmental biology and from the evolutionary perspective.
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
Ethyl 3-aminobenzoate, 97%