- Estimation of the shielding ability of a tungsten functional paper for diagnostic x-rays and gamma rays.
Estimation of the shielding ability of a tungsten functional paper for diagnostic x-rays and gamma rays.
Journal of applied clinical medical physics (2017-06-29)
Hajime Monzen, Ikuo Kanno, Takahiro Fujimoto, Masahiro Hiraoka
PMID28656739
ABSTRACT
Tungsten functional paper (TFP) is a novel paper-based radiation-shielding material. We measured the shielding ability of TFP against x-rays and gamma rays. The TFP was supplied in 0.3-mm-thick sheets that contained 80% tungsten powder and 20% cellulose (C