Skip to Content
AT
EN
Products
Applications
Services
Resources
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Lab Automation
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Documents
Learning Center
New
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Home
Search Results
Low-affinity binding in
Low-affinity binding in
Molecular biology of the cell (2017-08-25)
Virginie Bondu, Chenyu Wu, Wenpeng Cao, Peter C Simons, Jennifer Gillette, Jieqing Zhu, Laurie Erb, X Frank Zhang, Tione Buranda
PMID
28835374
ABSTRACT
Pathogenic hantaviruses bind to the plexin-semaphorin-integrin (PSI) domain of inactive, β
Product Description
P0115
Sigma-Aldrich
PAC-1, ≥98% (HPLC)
Pricing
EHU156731
Sigma-Aldrich
MISSION
®
esiRNA, targeting human P2RY2
Pricing