Merck
  • Clenbuterol Induces Cell Cycle Arrest in C2C12 Myoblasts by Delaying p27 Degradation through β-arrestin 2 Signaling.
Clenbuterol Induces Cell Cycle Arrest in C2C12 Myoblasts by Delaying p27 Degradation through β-arrestin 2 Signaling.
International journal of biological sciences (2017-11-07)
Min Chen, Chuncheng Liu, Meng Wang, Hong Wang, Kuo Zhang, Yu Zheng, Zhengquan Yu, Xiangdong Li, Wei Guo, Ning Li, Qingyong Meng
ABSTRACT
β
Sigma-Aldrich
MISSION® esiRNA, targeting human CDKN1B
Sigma-Aldrich
Monoclonal Anti-BrdU antibody produced in mouse, clone BU-33, ascites fluid, Immunohistology Grade