Merck
  • Straightforward analytical method to determine opium alkaloids in poppy seeds and bakery products.
Straightforward analytical method to determine opium alkaloids in poppy seeds and bakery products.
Food chemistry (2017-10-19)
Patricia López, Diana P K H Pereboom-de Fauw, Patrick P J Mulder, Martien Spanjer, Joyce de Stoppelaar, Hans G J Mol, Monique de Nijs
ABSTRACT
A straightforward method to determine the content of six opium alkaloids (morphine, codeine, thebaine, noscapine, papaverine and narceine) in poppy seeds and bakery products was developed and validated down to a limit of quantification (LOQ) of 0.1mg/kg. The method was based on extraction with acetonitrile/water/formic acid, ten-fold dilution and analysis by LC-MS/MS using a pH 10 carbonate buffer. The method was applied for the analysis of 41 samples collected in 2015 in the Netherlands and Germany. All samples contained morphine ranging from 0.2 to 240mg/kg. The levels of codeine and thebaine ranged from below LOQ to 348mg/kg and from below LOQ to 106mg/kg, respectively. Sixty percent of the samples exceeded the guidance reference value of 4mg/kg of morphine set by BfR in Germany, whereas 25% of the samples did not comply with the limits set for morphine, codeine, thebaine and noscapine by Hungarian legislation.
Product Description
Supelco
Noscapine hydrochloride hydrate, analytical standard
Pricing and availability is not currently available.