Merck
Temporal Control of Mammalian Cortical Neurogenesis by m6A Methylation.
Cell (2017-10-03)
Ki-Jun Yoon, Francisca Rojas Ringeling, Caroline Vissers, Fadi Jacob, Michael Pokrass, Dennisse Jimenez-Cyrus, Yijing Su, Nam-Shik Kim, Yunhua Zhu, Lily Zheng, Sunghan Kim, Xinyuan Wang, Louis C Doré, Peng Jin, Sergi Regot, Xiaoxi Zhuang, Stefan Canzar, Chuan He, Guo-Li Ming, Hongjun Song
ABSTRACT
N6-methyladenosine (m6A), installed by the Mettl3/Mettl14 methyltransferase complex, is the most prevalent internal mRNA modification. Whether m6A regulates mammalian brain development is unknown. Here, we show that m6A depletion by Mettl14 knockout in embryonic mouse brains prolongs the cell cycle of radial glia cells and extends cortical neurogenesis into postnatal stages. m6A depletion by Mettl3 knockdown also leads to a prolonged cell cycle and maintenance of radial glia cells. m6A sequencing of embryonic mouse cortex reveals enrichment of mRNAs related to transcription factors, neurogenesis, the cell cycle, and neuronal differentiation, and m6A tagging promotes their decay. Further analysis uncovers previously unappreciated transcriptional prepatterning in cortical neural stem cells. m6A signaling also regulates human cortical neurogenesis in forebrain organoids. Comparison of m6A-mRNA landscapes between mouse and human cortical neurogenesis reveals enrichment of human-specific m6A tagging of transcripts related to brain-disorder risk genes. Our study identifies an epitranscriptomic mechanism in heightened transcriptional coordination during mammalian cortical neurogenesis.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
(1R,2S,5R)-(−)-Menthol, ReagentPlus®, 99%
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
Methylene blue, certified by the BSC
Sigma-Aldrich
N6-Methyladenosine 5′-monophosphate sodium salt, ≥97% (HPLC)
Sigma-Aldrich
4-Thiouridine, ≥98%
Sigma-Aldrich
Anti-β-Tubulin III antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution